Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ggps1
- Entrez Gene ID
-
14593
- Gene Name
- geranylgeranyl diphosphate synthase 1
- Gene Aliases
-
1810026C22Rik, 9530089B04Rik, AI843169, C79210
- Chromosome Location
-
Chr.13: 14049793 - 14063734 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK011980.1 |
- |
2 - 3 |
339 |
99 |
View Full Product Details