Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsfm
Entrez Gene ID
66399
Gene Name
Ts translation elongation factor, mitochondrial
Gene Aliases
2310050B20Rik, 9430024O13Rik, EF-TS, EF-Tsmt
Chromosome Location
Chr.10: 127022332 - 127030814 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025537.3 NP_079813.1 4 - 5 493 80
GenBank mRNA AJ277891.1 - 3 - 4 330 80
AK012240.1 - 4 - 5 484 80
AK018591.1 - 4 - 5 482 80
AK020437.1 - 4 - 5 478 80
AK169164.1 - 4 - 5 484 80
AK171656.1 - 4 - 5 490 80
AK172112.1 - 4 - 5 490 80
BC034286.1 - 4 - 5 472 80
BC051508.1 - 1 - 2 74 80
BC057904.1 - 4 - 5 493 80

View Full Product Details