Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chrdl2
Entrez Gene ID
69121
Gene Name
chordin-like 2
Gene Aliases
1810022C01Rik, Chl2
Chromosome Location
Chr.7: 100006404 - 100034728 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291320.1 NP_001278249.1 8 - 9 1053 70
NM_133709.3 NP_598470.3 8 - 9 1095 70
GenBank mRNA AF338222.1 - 8 - 9 999 70
AK007577.1 - 8 - 9 1058 70
BC019399.1 - 8 - 9 1060 70

View Full Product Details