Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Swap70
Entrez Gene ID
20947
Gene Name
SWA-70 protein
Gene Aliases
70kDa, AV235546
Chromosome Location
Chr.7: 110221703 - 110283506 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009302.3 NP_033328.3 11 - 12 1760 71
GenBank mRNA AF053974.1 - 11 - 12 1652 71
AK019882.1 - 11 - 12 1704 71
AK146773.1 - 11 - 12 1752 71
AK150621.1 - 5 - 6 732 71
AK163357.1 - 4 - 5 570 71
AK172990.1 - 11 - 12 1697 71

View Full Product Details