Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpys
Entrez Gene ID
64705
Gene Name
dihydropyrimidinase
Gene Aliases
1200017I10Rik, 1300004I01Rik, DHP, DHPase
Chromosome Location
Chr.15: 39768485 - 39857470 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164466.1 NP_001157938.1 2 - 3 523 77
NM_022722.3 NP_073559.3 2 - 3 523 77
GenBank mRNA AF227731.1 - 2 - 3 441 77
AF249296.1 - 2 - 3 510 77
AK004822.1 - 2 - 3 523 77
AK004899.2 - 2 - 3 520 77
BC037086.1 - 2 - 3 492 77

View Full Product Details