The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081459.2 | NP_001074928.1 | 2 - 3 | 1537 | 84 |
NM_001289538.1 | NP_001276467.1 | 2 - 3 | 1537 | 84 | |
NM_001289539.1 | NP_001276468.1 | 2 - 3 | 1160 | 84 | |
NM_001289540.1 | NP_001276469.1 | 3 - 4 | 1297 | 84 | |
NM_001289541.1 | NP_001276470.1 | 2 - 3 | 1160 | 84 | |
NM_001289542.1 | NP_001276471.1 | 2 - 3 | 1537 | 84 | |
NM_011363.3 | NP_035493.2 | 2 - 3 | 1537 | 84 | |
NR_110346.1 | 1 - 2 | 1025 | 84 | ||
GenBank mRNA | AF020526.1 | - | 2 - 3 | 1231 | 84 |
AF074329.1 | - | 1 - 2 | 942 | 84 | |
AF380422.1 | - | 2 - 3 | 1231 | 84 | |
AF421138.1 | - | 2 - 3 | 1231 | 84 | |
AF421139.1 | - | 2 - 3 | 1231 | 84 | |
AK148085.1 | - | 3 - 4 | 1264 | 84 | |
AK154655.1 | - | 2 - 3 | 1471 | 84 | |
AK168439.1 | - | 2 - 3 | 1132 | 84 | |
AK170444.1 | - | 2 - 3 | 1511 | 84 | |
AK173145.1 | - | 1 - 2 | 684 | 84 | |
BC011422.1 | - | 1 - 2 | 999 | 84 | |
BC051978.1 | - | 2 - 3 | 1080 | 84 |