The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081459.2 | NP_001074928.1 | 5 - 6 | 1906 | 65 |
NM_001289538.1 | NP_001276467.1 | 5 - 6 | 1906 | 65 | |
NM_001289539.1 | NP_001276468.1 | 5 - 6 | 1529 | 65 | |
NM_001289540.1 | NP_001276469.1 | 6 - 7 | 1666 | 65 | |
NM_001289541.1 | NP_001276470.1 | 5 - 6 | 1529 | 65 | |
NM_001289542.1 | NP_001276471.1 | 5 - 6 | 1906 | 65 | |
NM_011363.3 | NP_035493.2 | 5 - 6 | 1906 | 65 | |
GenBank mRNA | AF020526.1 | - | 5 - 6 | 1600 | 65 |
AF380422.1 | - | 5 - 6 | 1600 | 65 | |
AF421138.1 | - | 5 - 6 | 1600 | 65 | |
AF421139.1 | - | 5 - 6 | 1600 | 65 | |
AK148085.1 | - | 6 - 7 | 1633 | 65 | |
AK154655.1 | - | 5 - 6 | 1840 | 65 | |
AK168439.1 | - | 5 - 6 | 1501 | 65 | |
AK170444.1 | - | 5 - 6 | 1880 | 65 | |
AK171328.1 | - | 5 - 6 | 1914 | 65 | |
BC051978.1 | - | 5 - 6 | 1449 | 65 |