The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163664.1 | NP_001157136.1 | 16 - 17 | 809 | 97 |
NM_001163666.1 | NP_001157138.1 | 14 - 15 | 782 | 97 | |
NM_001163667.1 | NP_001157139.1 | 15 - 16 | 797 | 97 | |
NM_001163668.1 | NP_001157140.1 | 14 - 15 | 758 | 97 | |
NM_001163669.1 | NP_001157141.1 | 15 - 16 | 770 | 97 | |
NM_001163670.1 | NP_001157142.1 | 12 - 13 | 725 | 97 | |
NM_011620.3 | NP_035750.2 | 13 - 14 | 737 | 97 | |
GenBank mRNA | AI037743.1 | - | 2 - 3 | 132 | 97 |
AI322331.1 | - | 2 - 3 | 111 | 97 | |
AK003147.1 | - | 12 - 13 | 692 | 97 | |
CK791812.1 | - | 5 - 6 | 447 | 97 | |
L48989.1 | - | 10 - 11 | 628 | 97 | |
L48993.1 | - | 12 - 13 | 661 | 97 | |
L49018.1 | - | 10 - 11 | 655 | 97 | |
L49466.1 | - | 15 - 16 | 797 | 97 | |
L49467.1 | - | 16 - 17 | 809 | 97 | |
L49468.1 | - | 14 - 15 | 782 | 97 | |
L49470.1 | - | 15 - 16 | 770 | 97 | |
L49471.1 | - | 14 - 15 | 779 | 97 | |
L49472.1 | - | 14 - 15 | 758 | 97 |