Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptdss2
Entrez Gene ID
27388
Gene Name
phosphatidylserine synthase 2
Gene Aliases
AI481134, AI988017, PSS2
Chromosome Location
Chr.7: 141131286 - 141156158 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013782.4 NP_038810.2 6 - 7 674 64
GenBank mRNA AF099053.1 - 6 - 7 558 64
AK021249.1 - 6 - 7 639 64
AK031906.1 - 6 - 7 660 64
AK032686.1 - 6 - 7 639 64
AK036368.1 - 6 - 7 675 64
AK048026.1 - 6 - 7 657 64
AK081275.1 - 6 - 7 641 64
AK137415.1 - - 634 64
BC009013.1 - 4 - 5 341 64
BC053517.1 - 6 - 7 611 64

View Full Product Details