Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptdss2
Entrez Gene ID
27388
Gene Name
phosphatidylserine synthase 2
Gene Aliases
AI481134, AI988017, PSS2
Chromosome Location
Chr.7: 141131286 - 141156158 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013782.4 NP_038810.2 9 - 10 1018 82
GenBank mRNA AF099053.1 - 9 - 10 902 82
AK036368.1 - 9 - 10 1019 82
AK048026.1 - 9 - 10 1001 82
AK081275.1 - 9 - 10 985 82
AK137415.1 - - 979 82
BC009013.1 - 7 - 8 685 82
BC053517.1 - 9 - 10 955 82

View Full Product Details