The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145427.2 | NP_663402.2 | 1 - 2 | 348 | 65 |
GenBank mRNA | AK078500.1 | - | 1 - 2 | 293 | 65 |
BC013607.1 | - | 1 - 2 | 254 | 65 |