Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gga2
Entrez Gene ID
74105
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 2
Gene Aliases
1200007E24Rik, mKIAA1080
Chromosome Location
Chr.7: 121986722 - 122021198 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028758.2 NP_083034.1 14 - 15 1528 66
GenBank mRNA AK004632.1 - 14 - 15 1500 66
AK034612.1 - 11 - 12 1171 66
AK129280.1 - 14 - 15 1487 66
AK143691.1 - 14 - 15 1516 66
AK149798.1 - 14 - 15 1518 66
AK154904.1 - 14 - 15 1503 66
AK159953.1 - 14 - 15 1518 66
BC057377.1 - 7 - 8 1031 66
BC062933.1 - 14 - 15 1497 66

View Full Product Details