Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Srebf1
Entrez Gene ID
20787
Gene Name
sterol regulatory element binding transcription factor 1
Gene Aliases
ADD-1, ADD1, D630008H06, SREBP-1, SREBP-1a, SREBP-1c, SREBP1, SREBP1c, bHLHd1
Chromosome Location
Chr.11: 60199089 - 60222686 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011480.3 NP_035610.1 15 - 16 2761 101
GenBank mRNA AF374266.1 - 14 - 15 2525 101
AK052628.1 - 15 - 16 2762 101
AK150052.1 - 15 - 16 2750 101
AK154424.1 - 15 - 16 2729 101
AK158269.1 - 8 - 9 1310 101
AK169607.1 - 15 - 16 2762 101
AK205046.1 - 1 - 2 75 101
BC006051.1 - 12 - 13 1897 101
BC056922.1 - 15 - 16 2743 101

View Full Product Details