The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039092.3 | NP_001034181.1 | 8 - 9 | 1010 | 90 |
NM_001039093.1 | NP_001034182.1 | 8 - 9 | 1010 | 90 | |
NM_153080.2 | NP_694720.2 | 8 - 9 | 1010 | 90 | |
GenBank mRNA | AF467887.1 | - | 8 - 9 | 990 | 90 |
AK043095.1 | - | 8 - 9 | 990 | 90 | |
AK054531.1 | - | 8 - 9 | 978 | 90 | |
AK166040.1 | - | 8 - 9 | 1010 | 90 | |
BC062947.1 | - | 8 - 9 | 974 | 90 | |
BC127266.1 | - | 8 - 9 | 976 | 90 |