Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kif22
Entrez Gene ID
110033
Gene Name
kinesin family member 22
Gene Aliases
AU021460, C81217, Kid, Kif22a
Chromosome Location
Chr.7: 127027729 - 127042466 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145588.1 NP_663563.1 12 - 13 1897 55
GenBank mRNA AK075922.1 - 12 - 13 1888 55
AK084310.1 - 12 - 13 1886 55
AK088033.1 - 9 - 10 1473 55
BC003427.1 - 12 - 13 1897 55

View Full Product Details