Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ebf3
Entrez Gene ID
13593
Gene Name
early B cell factor 3
Gene Aliases
3110018A08Rik, O/E-2, mKIAA4201
Chromosome Location
Chr.7: 137193671 - 137322810 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113414.1 NP_001106885.1 6 - 7 1077 81
NM_001113415.1 NP_001106886.1 6 - 7 1077 81
NM_010096.3 NP_034226.1 6 - 7 1077 81
GenBank mRNA AK220542.1 - 6 - 7 1077 81
BC067018.1 - 7 - 8 832 81
U82442.1 - 5 - 6 374 81
U92702.1 - 6 - 7 618 81
U92704.1 - 6 - 7 618 81

View Full Product Details