The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113368.1 | NP_001106839.1 | 5 - 6 | 1482 | 89 |
NM_001113369.1 | NP_001106840.1 | 3 - 4 | 942 | 89 | |
NM_007543.4 | NP_031569.1 | 3 - 4 | 942 | 89 | |
GenBank mRNA | AB500065.1 | - | 3 - 4 | 767 | 89 |
AB731450.1 | - | 3 - 4 | 702 | 89 | |
AF101164.1 | - | 3 - 4 | 800 | 89 | |
BC024320.1 | - | 3 - 4 | 751 | 89 | |
X76085.1 | - | 3 - 4 | 800 | 89 |