Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vps11
Entrez Gene ID
71732
Gene Name
vacuolar protein sorting 11 (yeast)
Gene Aliases
1200011A11Rik
Chromosome Location
Chr.9: 44347881 - 44361675 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027889.1 NP_082165.1 5 - 6 927 66
GenBank mRNA AK004695.1 - 5 - 6 902 66
AK032484.1 - 5 - 6 892 66
AK037658.1 - 5 - 6 901 66
AK047228.1 - 5 - 6 735 66
AK090030.1 - 5 - 6 908 66
AK153963.1 - 5 - 6 900 66
AK162897.1 - 5 - 6 902 66
BC029004.1 - 5 - 6 927 66

View Full Product Details