Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cfd
Entrez Gene ID
11537
Gene Name
complement factor D (adipsin)
Gene Aliases
Adn, DF
Chromosome Location
Chr.10: 79890853 - 79892660 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291915.1 NP_001278844.1 3 - 4 431 110
NM_013459.3 NP_038487.1 3 - 4 434 110
GenBank mRNA AK143817.1 - 3 - 4 378 110
BC138778.1 - 3 - 4 434 110
BC138779.1 - 3 - 4 434 110
BC145412.1 - 3 - 4 431 110
DV051476.1 - 3 - 4 379 110
M11768.1 - 3 - 4 375 110

View Full Product Details