Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem259
Entrez Gene ID
216157
Gene Name
transmembrane protein 259
Gene Aliases
AC087114.7gm2, ORF61, R32184_3, mFLJ00277
Chromosome Location
Chr.10: 79977120 - 79984330 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003949.3 NP_001003949.2 8 - 9 1253 74
GenBank mRNA AY096036.1 - 8 - 9 963 74
BC005494.1 - 7 - 8 776 74
BC052787.1 - 7 - 8 795 74
BC100297.1 - 5 - 6 390 74

View Full Product Details