Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pald1
Entrez Gene ID
27355
Gene Name
phosphatase domain containing, paladin 1
Gene Aliases
MMPAL, Pald, mKIAA1274
Chromosome Location
Chr.10: 61319656 - 61383533 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013753.2 NP_038781.2 12 - 13 1718 92
GenBank mRNA AK088577.1 - 12 - 13 1554 92
AK146489.1 - 12 - 13 1663 92
BC137695.1 - 11 - 12 1482 92
BC144846.1 - 11 - 12 1491 92
BC144847.1 - 11 - 12 1488 92
X99384.1 - 11 - 12 1503 92

View Full Product Details