The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284328.1 | NP_001271257.1 | 1 - 2 | 349 | 94 |
NM_019666.2 | NP_062640.2 | 1 - 2 | 349 | 94 | |
NM_019796.5 | NP_062770.1 | 1 - 2 | 349 | 94 | |
GenBank mRNA | AK034845.1 | - | 1 - 2 | 161 | 94 |
AK076026.1 | - | 1 - 2 | 141 | 94 | |
AK077588.1 | - | 1 - 2 | 98 | 94 | |
BC041148.1 | - | 1 - 2 | 194 | 94 | |
BC050079.1 | - | 1 - 2 | 122 | 94 | |
BC055863.1 | - | 1 - 2 | 127 | 94 | |
BC058807.1 | - | 1 - 2 | 216 | 94 | |
BC080309.1 | - | 1 - 2 | 52 | 94 | |
BC108363.1 | - | 1 - 2 | 83 | 94 | |
BU505018.1 | - | 1 - 2 | 214 | 94 |