Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc51a
Entrez Gene ID
106407
Gene Name
solute carrier family 51, alpha subunit
Gene Aliases
AV001382, AW261577, D630035O19Rik, OSTalpha, Osta
Chromosome Location
Chr.16: 32475578 - 32487879 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145932.3 NP_666044.1 4 - 5 711 68
GenBank mRNA AK052725.1 - 4 - 5 712 68
AK147155.1 - - 531 68
BC024441.1 - 4 - 5 519 68
BC025912.1 - 4 - 5 522 68
BC031178.1 - 4 - 5 462 68

View Full Product Details