Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dgcr8
Entrez Gene ID
94223
Gene Name
DiGeorge syndrome critical region gene 8
Gene Aliases
D16H22S1742E, D16H22S788E, D16Wis2, Gy1, N41, Vo59c07, mir-1306
Chromosome Location
Chr.16: 18253948 - 18289252 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033324.2 NP_201581.2 5 - 6 1654 79
GenBank mRNA AB086857.1 - 5 - 6 1654 79
AK048056.1 - 4 - 5 1455 79
AK086233.1 - 4 - 5 1846 79
BC051013.1 - 3 - 4 1840 79
BC062919.1 - 5 - 6 1674 79
BC066118.1 - 5 - 6 1701 79

View Full Product Details