The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033324.2 | NP_201581.2 | 7 - 8 | 1951 | 64 |
GenBank mRNA | AB086857.1 | - | 7 - 8 | 1951 | 64 |
AK048056.1 | - | 6 - 7 | 1752 | 64 | |
AK086233.1 | - | 6 - 7 | 2143 | 64 | |
BC051013.1 | - | 5 - 6 | 2137 | 64 | |
BC062919.1 | - | 7 - 8 | 1971 | 64 | |
BC066118.1 | - | 7 - 8 | 1998 | 64 |