The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289813.1 | NP_001276742.1 | 2 - 3 | 363 | 57 |
NM_001289814.1 | NP_001276743.1 | 2 - 3 | 363 | 57 | |
NM_010047.4 | NP_034177.1 | 3 - 4 | 560 | 57 | |
GenBank mRNA | AF021031.1 | - | 2 - 3 | 264 | 57 |
AK019346.1 | - | 3 - 4 | 563 | 57 | |
AK160342.1 | - | 3 - 4 | 550 | 57 | |
BC048503.1 | - | 2 - 3 | 215 | 57 | |
BE945982.1 | - | 2 - 3 | 263 | 57 |