Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mier2
Entrez Gene ID
70427
Gene Name
mesoderm induction early response 1, family member 2
Gene Aliases
2700087H15Rik, mKIAA1193
Chromosome Location
Chr.10: 79540245 - 79555631 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027422.2 NP_081698.2 5 - 6 493 66
GenBank mRNA AK154672.1 - 5 - 6 510 66
AK173128.1 - 4 - 5 649 66
BC067013.1 - 5 - 6 493 66

View Full Product Details