Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sgta
Entrez Gene ID
52551
Gene Name
small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha
Gene Aliases
5330427H01Rik, AI194281, D10Ertd190e, Sgt, Stg
Chromosome Location
Chr.10: 81044071 - 81060232 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024499.1 NP_078775.1 3 - 4 270 86
GenBank mRNA AK079168.1 - 3 - 4 280 86
BC003836.1 - 3 - 4 270 86

View Full Product Details