The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025616.3 | NP_079892.1 | 10 - 11 | 972 | 88 |
GenBank mRNA | AK008340.1 | - | 10 - 11 | 973 | 88 |
AK008382.1 | - | 10 - 11 | 969 | 88 | |
AK010832.1 | - | 10 - 11 | 938 | 88 | |
AK012977.1 | - | 10 - 11 | 964 | 88 | |
AK088074.1 | - | 10 - 11 | 963 | 88 | |
AK167473.1 | - | 10 - 11 | 975 | 88 | |
AY551342.1 | - | 10 - 11 | 958 | 88 | |
BC010303.1 | - | 10 - 11 | 947 | 88 | |
BC025844.1 | - | 4 - 5 | 453 | 88 | |
BC037056.1 | - | 10 - 11 | 974 | 88 |