Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pou2f2
Entrez Gene ID
18987
Gene Name
POU domain, class 2, transcription factor 2
Gene Aliases
Oct-2, Oct2a, Oct2b, Otf-2, Otf2
Chromosome Location
Chr.7: 25091114 - 25179992 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163554.1 NP_001157026.1 9 - 10 765 61
NM_001163555.1 NP_001157027.1 9 - 10 831 61
NM_001163556.1 NP_001157028.1 9 - 10 765 61
NM_011138.2 NP_035268.2 9 - 10 813 61
GenBank mRNA AK156322.1 - 9 - 10 778 61
AY746973.1 - 9 - 10 793 61
AY746974.1 - 9 - 10 811 61
BC050258.1 - 9 - 10 778 61
BC104488.1 - 9 - 10 765 61
BC105647.1 - 9 - 10 765 61
BC105920.1 - 9 - 10 831 61
BC105921.1 - 9 - 10 765 61
S55236.1 - 2 - 3 270 61
U89413.1 - 1 - 2 80 61
X51961.1 - 1 - 2 151 61
X53654.1 - - 973 61
X57089.1 - 2 - 3 270 61
X57936.1 - 9 - 10 765 61
X57937.1 - 9 - 10 813 61
X57938.1 - 9 - 10 831 61
X57939.1 - 9 - 10 765 61
X57940.1 - 9 - 10 765 61
X57941.1 - 8 - 9 648 61

View Full Product Details