Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xrcc1
Entrez Gene ID
22594
Gene Name
X-ray repair complementing defective repair in Chinese hamster cells 1
Gene Aliases
Xrcc-1
Chromosome Location
Chr.7: 24546300 - 24573438 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009532.4 NP_033558.3 15 - 16 1867 67
GenBank mRNA AK155376.1 - 15 - 16 1844 67
AK165569.1 - 15 - 16 1822 67
AK168334.1 - 15 - 16 1817 67
AK168727.1 - 15 - 16 1821 67
AU020161.1 - 2 - 3 67
BC055900.1 - 15 - 16 1843 67
BC085281.1 - 17 - 18 2249 67
U02887.1 - 15 - 16 1831 67

View Full Product Details