Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adarb1
Entrez Gene ID
110532
Gene Name
adenosine deaminase, RNA-specific, B1
Gene Aliases
1700057H01Rik, AW124433, AW558573, Adar2, BB220382, D10Bwg0447e, Red1
Chromosome Location
Chr.10: 77290727 - 77418273 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024837.2 NP_001020008.1 10 - 11 2354 72
NM_130895.3 NP_570965.2 10 - 11 2324 72
NR_004429.1 10 - 11 2401 72
NR_021486.1 10 - 11 2371 72
GenBank mRNA AF291049.1 - 9 - 10 2159 72
AF403106.2 - 10 - 11 2276 72
AF403107.2 - 10 - 11 2306 72
AF403108.2 - 10 - 11 2323 72
AF403109.2 - 10 - 11 2353 72
AK139004.1 - 1 - 2 156 72
AK141777.1 - 10 - 11 2326 72
AK147298.1 - 11 - 12 2433 72
AY162454.1 - 11 - 12 2349 72
BC040200.1 - 6 - 7 909 72

View Full Product Details