The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162846.1 | NP_001156318.1 | 2 - 3 | 335 | 108 |
NM_178621.4 | NP_848736.2 | 2 - 3 | 485 | 108 | |
GenBank mRNA | AK014946.1 | - | 1 - 2 | 214 | 108 |
AK029522.1 | - | 2 - 3 | 472 | 108 | |
AK032166.1 | - | 2 - 3 | 486 | 108 | |
AK034448.1 | - | 2 - 3 | 466 | 107 | |
AK083011.1 | - | 2 - 3 | 486 | 108 | |
BC037618.1 | - | 2 - 3 | 473 | 108 | |
BC124999.1 | - | 2 - 3 | 283 | 108 |