Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cgnl1
Entrez Gene ID
68178
Gene Name
cingulin-like 1
Gene Aliases
4933421H10Rik, 9930020M10Rik, AI503810, Jacop
Chromosome Location
Chr.9: 71626507 - 71809300 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK173252.1 - 7 - 8 1475 82
BC139006.1 - 8 - 9 2619 82

View Full Product Details