The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271843.1 | NP_001258772.1 | 5 - 6 | 917 | 81 |
NM_022026.3 | NP_071309.1 | 5 - 6 | 917 | 81 | |
NR_073483.1 | 6 - 7 | 1067 | 81 | ||
GenBank mRNA | AB037180.1 | - | 5 - 6 | 714 | 81 |
AK050042.1 | - | 5 - 6 | 917 | 81 | |
AK154151.1 | - | 5 - 6 | 921 | 81 | |
BC024105.1 | - | 6 - 7 | 1038 | 81 | |
CT010174.1 | - | 5 - 6 | 711 | 81 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK050325.1 | - | 1973 | Chr. - - [-] on Build | 81 |