The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010348.3 | NP_034478.1 | 7 - 8 | 1659 | 67 |
NM_146072.4 | NP_666184.2 | 7 - 8 | 1659 | 67 | |
GenBank mRNA | AK042635.1 | - | 7 - 8 | 1660 | 67 |
AK043542.1 | - | 7 - 8 | 1639 | 67 | |
AK043737.1 | - | 7 - 8 | 1623 | 67 | |
AK044250.1 | - | 7 - 8 | 1618 | 67 | |
AK044420.1 | - | 7 - 8 | 1614 | 67 | |
AK046122.1 | - | 7 - 8 | 1666 | 67 | |
AK048634.1 | - | 7 - 8 | 1630 | 67 | |
AK133667.1 | - | 7 - 8 | 1645 | 67 | |
AK200858.1 | - | 1 - 2 | 154 | 67 | |
BC031822.1 | - | 6 - 7 | 1081 | 67 | |
BC049275.1 | - | 6 - 7 | 1081 | 67 | |
X66118.1 | - | 7 - 8 | 1645 | 67 |