Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ttk
Entrez Gene ID
22137
Gene Name
Ttk protein kinase
Gene Aliases
AL022661, Esk1, Mps1, PYT, esk
Chromosome Location
Chr.9: 83834651 - 83872390 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110265.1 NP_001103735.1 15 - 16 1777 152
NM_001284272.1 NP_001271201.1 15 - 16 1777 152
NM_009445.2 NP_033471.2 15 - 16 1777 152
GenBank mRNA AK041487.1 - 15 - 16 1748 152
AK164093.1 - 15 - 16 1776 152
AX775731.1 - 15 - 16 1743 152
BC058851.1 - 15 - 16 1763 152
DV659530.1 - 2 - 3 203 152
M86377.1 - 16 - 17 1803 152

View Full Product Details