Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trpc6
Entrez Gene ID
22068
Gene Name
transient receptor potential cation channel, subfamily C, member 6
Gene Aliases
AV025995, LLHWJM002, LLHWJM003, LLHWJM004, TRP-6, Trrp6, mtrp6
Chromosome Location
Chr.9: 8543932 - 8680752 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282086.1 NP_001269015.1 10 - 11 2614 130
NM_013838.2 NP_038866.2 11 - 12 2848 130
GenBank mRNA AF057748.1 - 11 - 12 2417 130
AK133864.1 - 10 - 11 2514 130
AK137447.1 - 10 - 11 2076 130
BC141131.1 - 11 - 12 2719 130
U49069.1 - 11 - 12 2848 130

View Full Product Details