The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145610.2 | NP_663585.2 | 10 - 11 | 1325 | 127 |
GenBank mRNA | AK039027.1 | - | 10 - 11 | 1230 | 127 |
AK171672.1 | - | 6 - 7 | 991 | 127 | |
AK172615.1 | - | 6 - 7 | 991 | 127 | |
BB749945.1 | - | 1 - 2 | 90 | 127 | |
BC014688.1 | - | 10 - 11 | 1243 | 127 | |
BC059277.1 | - | 8 - 9 | 1489 | 127 |