The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025401.3 | NP_079677.1 | 3 - 4 | 1730 | 63 |
GenBank mRNA | AK002730.1 | - | 3 - 4 | 239 | 63 |
AK003992.1 | - | 3 - 4 | 246 | 63 | |
AK007726.1 | - | 3 - 4 | 211 | 63 | |
AK009854.1 | - | 3 - 4 | 246 | 63 | |
AK012803.1 | - | 3 - 4 | 228 | 63 | |
AK051149.1 | - | 3 - 4 | 1731 | 63 | |
AK154036.1 | - | 2 - 3 | 2322 | 63 | |
BC028498.1 | - | 3 - 4 | 200 | 63 |