The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253897.1 | NP_001240826.1 | 3 - 4 | 379 | 90 |
NM_001253898.1 | NP_001240827.1 | 2 - 3 | 201 | 90 | |
NM_012029.2 | NP_036159.1 | 3 - 4 | 387 | 90 | |
GenBank mRNA | AF182510.1 | - | - | 400 | 90 |
AK007564.1 | - | 3 - 4 | 361 | 90 | |
AK012835.1 | - | 3 - 4 | 359 | 90 | |
BC004583.1 | - | 2 - 3 | 188 | 90 | |
BC016424.1 | - | 3 - 4 | 327 | 90 | |
BY123162.1 | - | 2 - 3 | 181 | 90 |