The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010019.3 | NP_034149.2 | 5 - 6 | 704 | 92 |
GenBank mRNA | AF052942.1 | - | 3 - 4 | 384 | 92 |
AK004350.1 | - | 5 - 6 | 677 | 92 | |
BC022165.1 | - | 5 - 6 | 673 | 92 | |
BX527735.1 | - | 1 - 2 | 88 | 92 | |
GU109477.1 | - | 5 - 6 | 704 | 92 |