The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028181.5 | NP_082457.3 | 8 - 9 | 2066 | 78 |
GenBank mRNA | AK076325.2 | - | 8 - 9 | 1993 | 78 |