Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dazap1
Entrez Gene ID
70248
Gene Name
DAZ associated protein 1
Gene Aliases
2410042M16Rik, mPrrp
Chromosome Location
Chr.10: 80261471 - 80288413 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122604.1 NP_001116076.1 5 - 6 576 72
NM_001122605.1 NP_001116077.1 5 - 6 573 72
NM_133188.2 NP_573451.2 5 - 6 573 72
GenBank mRNA AF225910.1 - 5 - 6 467 72
AK148023.1 - 5 - 6 451 72
AK152314.1 - 5 - 6 543 72
AK152345.1 - 5 - 6 543 72
BC049355.1 - 5 - 6 493 72
BC058775.1 - 5 - 6 487 72

View Full Product Details