Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fdxacb1
Entrez Gene ID
382137
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Gene Aliases
D630004A14Rik
Chromosome Location
Chr.9: 50768238 - 50772670 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198675.2 NP_941077.2 4 - 5 736 76
GenBank mRNA AK035237.1 - 2 - 3 1286 76
AK085276.1 - 4 - 5 657 76
AK135025.1 - - 739 76
BC098240.1 - 2 - 3 333 76
BC116265.2 - 2 - 3 1078 76

View Full Product Details