The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025548.3 | NP_079824.2 | 5 - 6 | 958 | 68 |
GenBank mRNA | AK002316.1 | - | 5 - 6 | 699 | 68 |
AK003655.2 | - | 5 - 6 | 704 | 68 | |
AK010438.1 | - | 5 - 6 | 715 | 68 | |
BC010684.1 | - | 5 - 6 | 691 | 68 |