The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205085.1 | NP_001192014.1 | 4 - 5 | 1110 | 68 |
NM_020496.3 | NP_065242.1 | 4 - 5 | 1110 | 68 | |
NM_194263.2 | NP_919239.1 | 4 - 5 | 1110 | 68 | |
GenBank mRNA | AF260557.1 | - | 4 - 5 | 1115 | 68 |
AF306667.1 | - | 4 - 5 | 1048 | 68 | |
AJ277486.1 | - | 4 - 5 | 652 | 68 | |
AK052155.1 | - | 4 - 5 | 1092 | 68 | |
AK052389.1 | - | 4 - 5 | 1093 | 68 | |
AK141251.1 | - | 4 - 5 | 880 | 68 | |
BY385297.1 | - | 2 - 3 | 141 | 68 |