Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lemd3
Entrez Gene ID
380664
Gene Name
LEM domain containing 3
Gene Aliases
AI316861, Man1
Chromosome Location
Chr.10: 120923411 - 120979949 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081193.2 NP_001074662.2 9 - 10 2326 137
GenBank mRNA BC082610.1 - 10 - 11 1090 137

View Full Product Details