Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lemd3
Entrez Gene ID
380664
Gene Name
LEM domain containing 3
Gene Aliases
AI316861, Man1
Chromosome Location
Chr.10: 120923411 - 120979949 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081193.2 NP_001074662.2 12 - 13 2597 106
GenBank mRNA AK140538.1 - 12 - 13 1080 106
BC082610.1 - 13 - 14 1360 106

View Full Product Details