Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppef1
Entrez Gene ID
237178
Gene Name
protein phosphatase with EF hand calcium-binding domain 1
Gene Aliases
PPEF-1
Chromosome Location
Chr.X: 160622419 - 160766000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011147.1 NP_035277.1 12 - 13 1523 116

View Full Product Details